Assay Details
Gene Symbol: Tob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169300.3 | NP_001162771.1 | 2 - 3 | 1290 | 74 | |
NM_001169301.2 | NP_001162772.1 | 2 - 3 | 1120 | 74 | ||
NM_132876.5 | NP_573104.1 | 2 - 3 | 1290 | 74 | ||
NM_206760.3 | NP_996483.2 | 2 - 3 | 1048 | 74 | ||
GenBank mRNA | BT010303.1 | - | 2 - 3 | 1129 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
