Assay Details
Gene Symbol: Tob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169300.3 | NP_001162771.1 | 3 - 4 | 1460 | 93 | |
NM_001169301.2 | NP_001162772.1 | 3 - 4 | 1290 | 93 | ||
NM_132876.5 | NP_573104.1 | 3 - 4 | 1460 | 93 | ||
GenBank mRNA | BT010303.1 | - | 3 - 4 | 1299 | 93 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
