Assay Details
| Gene Symbol: Dsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144731.3 | NP_001138203.1 | 4 - 5 | 1056 | 55 | |
| NM_001298393.1 | NP_001285322.1 | 6 - 7 | 1097 | 55 | ||
| NM_080715.4 | NP_542446.2 | 6 - 7 | 1690 | 55 | ||
| NM_167502.3 | NP_727959.2 | 5 - 6 | 1754 | 55 | ||
| NM_167503.3 | NP_727960.1 | 6 - 7 | 1169 | 55 | ||
| NM_167504.2 | NP_727961.1 | 6 - 7 | 1094 | 55 | ||
| NM_206762.3 | NP_996485.1 | 6 - 7 | 1238 | 55 | ||
| GenBank mRNA | AY060841.1 | - | 6 - 7 | 1081 | 55 | |
| BT126193.1 | - | 6 - 7 | 1096 | 55 | ||
More Information
Set Membership: |
|

Dsp1
Genomic Map