Assay Details
Gene Symbol: Fur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169302.2 | NP_001162773.1 | 9 - 10 | 3610 | 62 | |
NM_001169303.2 | NP_001162774.1 | 11 - 12 | 3487 | 62 | ||
NM_078644.4 | NP_523368.2 | 10 - 11 | 3559 | 62 | ||
NM_167506.3 | NP_727963.1 | 10 - 11 | 3568 | 62 | ||
NM_167507.3 | NP_727964.1 | 10 - 11 | 3668 | 62 | ||
NM_167508.2 | NP_727965.1 | 10 - 11 | 3602 | 62 | ||
NM_167509.3 | NP_727966.1 | 10 - 11 | 3662 | 62 | ||
NM_167510.3 | NP_727967.1 | 11 - 12 | 3779 | 62 | ||
NM_206763.2 | NP_996486.1 | 10 - 11 | 3546 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
