Assay Details
| Gene Symbol: Fur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169302.2 | NP_001162773.1 | 3 - 4 | 1140 | 61 | |
| NM_001169303.2 | NP_001162774.1 | 3 - 4 | 1140 | 61 | ||
| NM_078644.4 | NP_523368.2 | 3 - 4 | 1140 | 61 | ||
| NM_167506.3 | NP_727963.1 | 3 - 4 | 1140 | 61 | ||
| NM_167507.3 | NP_727964.1 | 3 - 4 | 1240 | 61 | ||
| NM_167508.2 | NP_727965.1 | 3 - 4 | 1174 | 61 | ||
| NM_167509.3 | NP_727966.1 | 3 - 4 | 1243 | 61 | ||
| NM_167510.3 | NP_727967.1 | 4 - 5 | 1360 | 61 | ||
| NM_206763.2 | NP_996486.1 | 3 - 4 | 1127 | 61 | ||
More Information
Set Membership: |
|

Fur2
Genomic Map