Assay Details
| Gene Symbol: Fur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169302.2 | NP_001162773.1 | 4 - 5 | 1392 | 71 | |
| NM_001169303.2 | NP_001162774.1 | 5 - 6 | 1320 | 71 | ||
| NM_078644.4 | NP_523368.2 | 4 - 5 | 1392 | 71 | ||
| NM_167509.3 | NP_727966.1 | 4 - 5 | 1495 | 71 | ||
| NM_167510.3 | NP_727967.1 | 5 - 6 | 1612 | 71 | ||
| NM_206763.2 | NP_996486.1 | 4 - 5 | 1379 | 71 | ||
More Information
Set Membership: |
|

Fur2
Genomic Map