Assay Details
| Gene Symbol: Fur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169302.2 | NP_001162773.1 | 7 - 8 | 3263 | 55 | |
| NM_001169303.2 | NP_001162774.1 | 9 - 10 | 3140 | 55 | ||
| NM_078644.4 | NP_523368.2 | 8 - 9 | 3212 | 55 | ||
| NM_167506.3 | NP_727963.1 | 8 - 9 | 3221 | 55 | ||
| NM_167507.3 | NP_727964.1 | 8 - 9 | 3321 | 55 | ||
| NM_167508.2 | NP_727965.1 | 8 - 9 | 3255 | 55 | ||
| NM_167509.3 | NP_727966.1 | 8 - 9 | 3315 | 55 | ||
| NM_167510.3 | NP_727967.1 | 9 - 10 | 3432 | 55 | ||
| NM_206763.2 | NP_996486.1 | 8 - 9 | 3199 | 55 | ||
More Information
Set Membership: |
|

Fur2
Genomic Map