Assay Details
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298579.1 | NP_001285508.1 | 2 - 3 | 317 | 73 | |
NM_134580.3 | NP_608424.1 | 2 - 3 | 440 | 73 | ||
NM_167726.2 | NP_728384.1 | 2 - 3 | 519 | 73 | ||
GenBank mRNA | AY058508.1 | - | 2 - 3 | 291 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
