Assay Details
Gene Symbol: CG15445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272827.1 | NP_001259756.1 | 3 - 4 | 864 | 72 | |
NM_134592.4 | NP_608436.4 | 3 - 4 | 858 | 72 | ||
NM_167729.2 | NP_728390.1 | 2 - 3 | 918 | 72 | ||
NM_167730.2 | NP_728391.3 | 3 - 4 | 869 | 72 | ||
NM_167731.2 | NP_728392.2 | 3 - 4 | 865 | 72 | ||
GenBank mRNA | BT004870.1 | - | 2 - 3 | 896 | 72 | |
BT128735.1 | - | 3 - 4 | 862 | 72 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
