Assay Details
| Gene Symbol: Tbc1d15-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272877.1 | NP_001259806.1 | 2 - 3 | 1345 | 91 | |
| NM_001272878.1 | NP_001259807.1 | 2 - 3 | 1118 | 91 | ||
| NM_134659.4 | NP_608503.1 | 2 - 3 | 1118 | 91 | ||
| GenBank mRNA | AY051746.1 | - | 2 - 3 | 1125 | 91 | |
More Information
Set Membership: |
|

Tbc1d15-17
Genomic Map