Assay Details
Gene Symbol: CG17646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134774.3 | NP_608618.1 | 9 - 10 | 2296 | 59 | |
NM_164436.2 | NP_722727.1 | 9 - 10 | 1874 | 59 | ||
GenBank mRNA | AY069801.1 | - | 9 - 10 | 2273 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
