Assay Details
Gene Symbol: Drp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273017.1 | NP_001259946.1 | 4 - 5 | 1524 | 67 | |
NM_134850.3 | NP_608694.2 | 4 - 5 | 1524 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
