Assay Details
| Gene Symbol: CG3277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134918.4 | NP_608762.3 | 3 - 4 | 1100 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG3277| Gene Symbol: CG3277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134918.4 | NP_608762.3 | 3 - 4 | 1100 | 62 | |
Set Membership: |
|