Assay Details
Gene Symbol: pix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140015.3 | NP_648272.1 | 2 - 3 | 319 | 61 | |
NM_168306.2 | NP_729435.1 | 1 - 2 | 280 | 61 | ||
GenBank mRNA | AY089644.1 | - | 2 - 3 | 308 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
