Assay Details
Gene Symbol: pix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140015.3 | NP_648272.1 | 3 - 4 | 933 | 66 | |
NM_168306.2 | NP_729435.1 | 2 - 3 | 894 | 66 | ||
GenBank mRNA | AY089644.1 | - | 3 - 4 | 922 | 66 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
