Assay Details
Gene Symbol: Galk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202145.1 | NP_001189074.1 | 4 - 5 | 771 | 59 | |
NM_140019.4 | NP_648276.1 | 4 - 5 | 789 | 59 | ||
NM_168307.3 | NP_729438.1 | 3 - 4 | 463 | 59 | ||
NM_168308.3 | NP_729439.1 | 3 - 4 | 524 | 59 | ||
GenBank mRNA | AY058337.1 | - | 4 - 5 | 763 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
