Assay Details
Gene Symbol: Doc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140026.3 | NP_648283.1 | 1 - 2 | 252 | 113 | |
GenBank mRNA | AB035412.1 | - | 1 - 2 | 243 | 113 | |
BT025226.1 | - | 1 - 2 | 250 | 113 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
