Assay Details
| Gene Symbol: Bet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140055.4 | NP_648312.3 | 1 - 2 | 144 | 63 | |
| GenBank mRNA | BT015980.1 | - | 1 - 2 | 130 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bet3| Gene Symbol: Bet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140055.4 | NP_648312.3 | 1 - 2 | 144 | 63 | |
| GenBank mRNA | BT015980.1 | - | 1 - 2 | 130 | 63 | |
Set Membership: |
|