Assay Details
| Gene Symbol: APP-BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274770.1 | NP_001261699.1 | 4 - 5 | 1267 | 68 | |
| NM_140178.4 | NP_648435.1 | 4 - 5 | 1484 | 68 | ||
| GenBank mRNA | BT050569.1 | - | 4 - 5 | 1374 | 68 | |
More Information
Set Membership: |
|

APP-BP1
Genomic Map