Assay Details
Gene Symbol: wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140188.4 | NP_648445.1 | 1 - 2 | 1494 | 58 | |
GenBank mRNA | AY060600.1 | - | 1 - 2 | 1481 | 58 | |
DQ305404.1 | - | 1 - 2 | 1510 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
