Assay Details
| Gene Symbol: Ube3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140195.4 | NP_648452.1 | 5 - 6 | 2286 | 80 | |
| GenBank mRNA | AY069489.1 | - | 5 - 6 | 2231 | 80 | |
More Information
Set Membership: |
|

Ube3a
Genomic Map