Assay Details
Gene Symbol: Pop2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382181.1 | NP_001369048.1 | 3 - 4 | 1090 | 62 | |
NM_140281.3 | NP_648538.1 | 3 - 4 | 1090 | 62 | ||
NM_168484.2 | NP_729775.1 | 4 - 5 | 1295 | 62 | ||
NM_168485.2 | NP_729776.1 | 4 - 5 | 1229 | 62 | ||
GenBank mRNA | AY047553.1 | - | 3 - 4 | 1041 | 62 | |
BT001837.1 | - | 4 - 5 | 1188 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
