Assay Details
Gene Symbol: CrzR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274826.1 | NP_001261755.1 | 4 - 5 | 1702 | 81 | |
NM_140314.4 | NP_648571.1 | 4 - 5 | 1702 | 81 | ||
GenBank mRNA | AY071408.1 | - | 4 - 5 | 1694 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
