Assay Details
Gene Symbol: Smyd4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140317.3 | NP_648574.1 | 3 - 4 | 1570 | 85 | |
GenBank mRNA | AY071300.1 | - | 3 - 4 | 1457 | 85 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
