Assay Details
| Gene Symbol: Atg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169962.2 | NP_001163433.1 | 3 - 4 | 1028 | 82 | |
| NM_140344.3 | NP_648601.1 | 3 - 4 | 1028 | 82 | ||
| GenBank mRNA | AY129445.1 | - | 3 - 4 | 997 | 82 | |
| BT021340.1 | - | 3 - 4 | 963 | 82 | ||
More Information
Set Membership: |
|

Atg1
Genomic Map