Assay Details
Gene Symbol: Atg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169962.2 | NP_001163433.1 | 6 - 7 | 1384 | 72 | |
NM_140344.3 | NP_648601.1 | 6 - 7 | 1384 | 72 | ||
GenBank mRNA | AY129445.1 | - | 6 - 7 | 1353 | 72 | |
BT021340.1 | - | 6 - 7 | 1319 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
