Assay Details
| Gene Symbol: Wbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144472.3 | NP_001137944.1 | 1 - 2 | 162 | 73 | |
| NM_001144473.3 | NP_001137945.1 | 1 - 2 | 162 | 73 | ||
| NM_001274856.2 | NP_001261785.1 | 1 - 2 | 162 | 73 | ||
| NM_001274857.2 | NP_001261786.1 | 1 - 2 | 162 | 73 | ||
| NM_001274858.2 | NP_001261787.1 | 1 - 2 | 162 | 73 | ||
| NM_140350.5 | NP_648607.3 | 1 - 2 | 162 | 73 | ||
| GenBank mRNA | AY060828.1 | - | 1 - 2 | 132 | 73 | |
| BT126101.1 | - | 1 - 2 | 124 | 73 | ||
More Information
Set Membership: |
|

Wbp2
Genomic Map