Assay Details
Gene Symbol: eIF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258978.3 | NP_001245907.1 | 4 - 5 | 1141 | 93 | |
NM_001273188.1 | NP_001260117.1 | 4 - 5 | 1297 | 93 | ||
NM_057247.5 | NP_476595.1 | 4 - 5 | 1141 | 93 | ||
NM_164668.3 | NP_723137.1 | 4 - 5 | 1141 | 93 | ||
NM_164669.3 | NP_723138.1 | 4 - 5 | 1297 | 93 | ||
NM_164670.4 | NP_723139.1 | 4 - 5 | 1528 | 93 | ||
GenBank mRNA | AF145621.1 | - | 4244 | 93 | ||
AY069283.1 | - | 4 - 5 | 1136 | 93 | ||
AY121623.1 | - | 2032 | 93 | |||
BT099924.1 | - | 4 - 5 | 1264 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
