Assay Details
| Gene Symbol: Cpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273199.1 | NP_001260128.1 | 5 - 6 | 1391 | 81 | |
| NM_001273200.1 | NP_001260129.1 | 5 - 6 | 1489 | 81 | ||
| NM_057810.4 | NP_477158.1 | 5 - 6 | 1549 | 81 | ||
| NM_164689.2 | NP_723173.1 | 3 - 4 | 967 | 81 | ||
| GenBank mRNA | AY052000.1 | - | 5 - 6 | 1529 | 81 | |
More Information
Set Membership: |
|

Cpr
Genomic Map