Assay Details
Gene Symbol: eya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258984.1 | NP_001245913.1 | 3 - 4 | 2086 | 74 | |
NM_078768.4 | NP_523492.1 | 4 - 5 | 2574 | 74 | ||
NM_164693.2 | NP_723188.1 | 4 - 5 | 2329 | 74 | ||
GenBank mRNA | AY047539.1 | - | 4 - 5 | 2538 | 74 | |
L08501.1 | - | 4 - 5 | 2495 | 74 | ||
L08502.1 | - | 4 - 5 | 2329 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
