Assay Details
Gene Symbol: AkhR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273220.1 | NP_001260149.1 | 4 - 5 | 1307 | 57 | |
NM_058039.4 | NP_477387.1 | 4 - 5 | 751 | 57 | ||
NM_164705.3 | NP_723206.1 | 5 - 6 | 906 | 57 | ||
NM_205917.3 | NP_995639.1 | 4 - 5 | 751 | 57 | ||
GenBank mRNA | BT031198.1 | - | 4 - 5 | 731 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
