Assay Details
Gene Symbol: AkhR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273220.1 | NP_001260149.1 | 5 - 6 | 1791 | 53 | |
NM_058039.4 | NP_477387.1 | 5 - 6 | 1235 | 53 | ||
NM_164705.3 | NP_723206.1 | 6 - 7 | 1390 | 53 | ||
NM_205917.3 | NP_995639.1 | 5 - 6 | 1235 | 53 | ||
GenBank mRNA | BT031198.1 | - | 5 - 6 | 1215 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
