Assay Details
Gene Symbol: sens-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298763.1 | NP_001285692.1 | 3 - 4 | 2336 | 89 | |
NM_164716.2 | NP_723223.1 | 3 - 4 | 1953 | 89 | ||
GenBank mRNA | BT133368.1 | - | 3 - 4 | 1953 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
