Assay Details
| Gene Symbol: Hrb27C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169426.2 | NP_001162897.1 | 2 - 3 | 777 | 75 | |
| NM_001169427.2 | NP_001162898.1 | 3 - 4 | 750 | 75 | ||
| NM_001258988.2 | NP_001245917.1 | 2 - 3 | 800 | 75 | ||
| NM_001258989.2 | NP_001245918.1 | 3 - 4 | 823 | 75 | ||
| NM_001258990.2 | NP_001245919.1 | 3 - 4 | 869 | 75 | ||
| NM_057521.6 | NP_476869.1 | 3 - 4 | 873 | 75 | ||
| NM_164720.4 | NP_723228.1 | 3 - 4 | 875 | 75 | ||
| NM_164721.3 | NP_723229.1 | 3 - 4 | 896 | 75 | ||
| GenBank mRNA | AY069699.1 | - | 2 - 3 | 788 | 75 | |
| AY128430.1 | - | 3 - 4 | 1054 | 75 | ||
| DQ062784.1 | - | 1 - 2 | 497 | 75 | ||
More Information
Set Membership: |
|

Hrb27C
Genomic Map