Assay Details
Gene Symbol: ninaC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273260.1 | NP_001260189.1 | 4 - 5 | 2234 | 119 | |
NM_001273261.1 | NP_001260190.1 | 4 - 5 | 2234 | 119 | ||
NM_078779.3 | NP_523503.2 | 4 - 5 | 2268 | 119 | ||
NM_164747.1 | NP_723271.1 | 4 - 5 | 2234 | 119 | ||
GenBank mRNA | BT150399.1 | - | 4 - 5 | 2252 | 119 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
