Assay Details
Gene Symbol: ninaC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273260.1 | NP_001260189.1 | 9 - 10 | 3341 | 66 | |
NM_001273261.1 | NP_001260190.1 | 9 - 10 | 3341 | 66 | ||
NM_078779.3 | NP_523503.2 | 9 - 10 | 3375 | 66 | ||
NM_164747.1 | NP_723271.1 | 9 - 10 | 3341 | 66 | ||
GenBank mRNA | BT150399.1 | - | 9 - 10 | 3359 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
