Assay Details
| Gene Symbol: PGAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273302.1 | NP_001260231.1 | 1 - 2 | 587 | 67 | |
| NM_135347.3 | NP_609191.1 | 1 - 2 | 516 | 67 | ||
| NM_164798.3 | NP_723359.1 | 1 - 2 | 516 | 67 | ||
| GenBank mRNA | AY094655.1 | - | 1 - 2 | 431 | 67 | |
More Information
Set Membership: |
|

PGAP5
Genomic Map