Assay Details
| Gene Symbol: Pvr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169438.2 | NP_001162909.1 | 6 - 7 | 2037 | 60 | |
| NM_001169439.2 | NP_001162910.1 | 7 - 8 | 1624 | 60 | ||
| NM_001169440.2 | NP_001162911.1 | 7 - 8 | 1624 | 60 | ||
| NM_001201791.2 | NP_001188720.1 | 6 - 7 | 1420 | 60 | ||
| NM_001201792.2 | NP_001188721.1 | 6 - 7 | 1293 | 60 | ||
| NM_001273305.1 | NP_001260234.1 | 7 - 8 | 1624 | 60 | ||
| NM_001273306.1 | NP_001260235.1 | 6 - 7 | 1417 | 60 | ||
| NM_078785.4 | NP_523509.2 | 6 - 7 | 1420 | 60 | ||
| NM_164801.3 | NP_723365.1 | 6 - 7 | 1420 | 60 | ||
| NM_175983.3 | NP_787997.2 | 6 - 7 | 1402 | 60 | ||
| NM_205925.3 | NP_995647.2 | 6 - 7 | 1417 | 60 | ||
| NM_205926.3 | NP_995648.1 | 6 - 7 | 1455 | 60 | ||
| NM_205927.3 | NP_995649.1 | 6 - 7 | 2037 | 60 | ||
| GenBank mRNA | BT050546.1 | - | 5 - 6 | 1169 | 60 | |
| BT133154.1 | - | 7 - 8 | 1593 | 60 | ||
More Information
Set Membership: |
|

Pvr
Genomic Map