Assay Details
| Gene Symbol: Pvr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169438.2 | NP_001162909.1 | 9 - 10 | 2838 | 92 | |
| NM_001169439.2 | NP_001162910.1 | 10 - 11 | 2425 | 92 | ||
| NM_001169440.2 | NP_001162911.1 | 10 - 11 | 2425 | 92 | ||
| NM_001201791.2 | NP_001188720.1 | 8 - 9 | 2194 | 92 | ||
| NM_001201792.2 | NP_001188721.1 | 9 - 10 | 2094 | 92 | ||
| NM_001273305.1 | NP_001260234.1 | 9 - 10 | 2398 | 92 | ||
| NM_001273306.1 | NP_001260235.1 | 8 - 9 | 2191 | 92 | ||
| NM_078785.4 | NP_523509.2 | 9 - 10 | 2221 | 92 | ||
| NM_164801.3 | NP_723365.1 | 9 - 10 | 2221 | 92 | ||
| NM_175983.3 | NP_787997.2 | 9 - 10 | 2203 | 92 | ||
| NM_205925.3 | NP_995647.2 | 8 - 9 | 2191 | 92 | ||
| NM_205926.3 | NP_995648.1 | 9 - 10 | 2256 | 92 | ||
| NM_205927.3 | NP_995649.1 | 9 - 10 | 2838 | 92 | ||
| GenBank mRNA | BT050546.1 | - | 8 - 9 | 1970 | 92 | |
| BT133154.1 | - | 10 - 11 | 2394 | 92 | ||
More Information
Set Membership: |
|

Pvr
Genomic Map