Assay Details
| Gene Symbol: Mtpalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135455.3 | NP_609299.1 | 2 - 3 | 981 | 59 | |
| NM_164860.2 | NP_723470.1 | 2 - 3 | 1132 | 59 | ||
| GenBank mRNA | AF181648.1 | - | 2 - 3 | 946 | 59 | |
More Information
Set Membership: |
|

Mtpalpha
Genomic Map