Assay Details
Gene Symbol: Trp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259031.2 | NP_001245960.1 | 2 - 3 | 1015 | 67 | |
NM_001298863.1 | NP_001285792.1 | 2 - 3 | 967 | 67 | ||
NM_057660.4 | NP_477008.1 | 2 - 3 | 967 | 67 | ||
NM_164885.3 | NP_723510.1 | 2 - 3 | 1015 | 67 | ||
GenBank mRNA | BT003520.1 | - | 2 - 3 | 965 | 67 | |
BT099837.1 | - | 2 - 3 | 1009 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
