Assay Details
Gene Symbol: Trp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057660.4 | NP_477008.1 | 3 - 4 | 1275 | 56 | |
NM_164885.3 | NP_723510.1 | 3 - 4 | 1323 | 56 | ||
GenBank mRNA | BT003520.1 | - | 3 - 4 | 1273 | 56 | |
BT099837.1 | - | 3 - 4 | 1317 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
