Assay Details
| Gene Symbol: Utx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201844.1 | NP_001188773.1 | 14 - 15 | 3307 | 71 | |
| NM_001273390.1 | NP_001260319.1 | 12 - 13 | 3038 | 71 | ||
| NM_135524.4 | NP_609368.3 | 14 - 15 | 3325 | 71 | ||
| NM_164907.4 | NP_723550.2 | 12 - 13 | 2860 | 71 | ||
| NM_164908.4 | NP_723551.2 | 12 - 13 | 2848 | 71 | ||
| GenBank mRNA | AY129437.1 | - | 14 - 15 | 3390 | 71 | |
| BT010089.1 | - | 14 - 15 | 3325 | 71 | ||
More Information
Set Membership: |
|

Utx
Genomic Map