Assay Details
| Gene Symbol: Utx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201844.1 | NP_001188773.1 | 8 - 9 | 1595 | 65 | |
| NM_001273390.1 | NP_001260319.1 | 6 - 7 | 1326 | 65 | ||
| NM_135524.4 | NP_609368.3 | 8 - 9 | 1598 | 65 | ||
| NM_164907.4 | NP_723550.2 | 6 - 7 | 1133 | 65 | ||
| NM_164908.4 | NP_723551.2 | 6 - 7 | 1121 | 65 | ||
| GenBank mRNA | AY129437.1 | - | 8 - 9 | 1663 | 65 | |
| BT010089.1 | - | 8 - 9 | 1598 | 65 | ||
More Information
Set Membership: |
|

Utx
Genomic Map