Assay Details
| Gene Symbol: SamDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057875.5 | NP_477223.2 | 2 - 3 | 246 | 108 | |
| GenBank mRNA | BT021946.1 | - | 2 - 3 | 232 | 108 | |
More Information
Set Membership: |
|

SamDC
Genomic Map