Assay Details
| Gene Symbol: Ge-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135642.3 | NP_609486.2 | 3 - 4 | 723 | 73 | |
| NM_164957.2 | NP_723654.1 | 2 - 3 | 806 | 73 | ||
| GenBank mRNA | BT004479.1 | - | 3 - 4 | 703 | 73 | |
More Information
Set Membership: |
|

Ge-1
Genomic Map