Assay Details
Gene Symbol: Ge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135642.3 | NP_609486.2 | 9 - 10 | 3598 | 74 | |
NM_164957.2 | NP_723654.1 | 8 - 9 | 3681 | 74 | ||
GenBank mRNA | BT004479.1 | - | 9 - 10 | 3578 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
