Assay Details
| Gene Symbol: Sox21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274899.1 | NP_001261828.1 | 4 - 5 | 1781 | 84 | |
| NM_001274900.1 | NP_001261829.1 | 4 - 5 | 1781 | 84 | ||
| NM_140438.3 | NP_648695.1 | 4 - 5 | 1228 | 84 | ||
| GenBank mRNA | AY058313.1 | - | 4 - 5 | 1228 | 84 | |
More Information
Set Membership: |
|

Sox21b
Genomic Map