Assay Details
Gene Symbol: nan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274904.1 | NP_001261833.1 | 2 - 3 | 852 | 61 | |
NM_140439.3 | NP_648696.2 | 2 - 3 | 852 | 61 | ||
GenBank mRNA | AY262004.1 | - | 2 - 3 | 853 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
