Assay Details
Gene Symbol: nan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274904.1 | NP_001261833.1 | 6 - 7 | 2025 | 66 | |
NM_140439.3 | NP_648696.2 | 6 - 7 | 2025 | 66 | ||
GenBank mRNA | AY262004.1 | - | 6 - 7 | 2026 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
